Learn More
Brij(R) C20, Spectrum™ Chemical
BR106, 9004-95-9, C16H33(OCH2CH2)20OH
Supplier:  Spectrum Chemical Mfg Cor BR1062.5KG
Quantity 2.5 kg
Packaging Amber Glass Bottle
 View more versions of this product
Catalog No. 18601066
Only null left
Description
Description
Spectrum™ Chemical Brij C20 is a non-ionic detergent for protein extraction and permeabilization of cells. It may be used in the preparation of yeast spheroplasts. It appears as a white, waxy solid.
Specifications
Chemical Identifiers
9004-95-9
1123.51
NLMKTBGFQGKQEV-UHFFFAOYSA-N
CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO
C56H114O21
MFCD00080892
3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxahexaheptacontan-1-ol
Specifications
Brij 58
100%
Amber Glass Bottle
MFCD00080892
CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO
1123.51
9004-95-9
2.5 kg
C56H114O21
NLMKTBGFQGKQEV-UHFFFAOYSA-N
3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxahexaheptacontan-1-ol
Ungraded
Product Suggestions
Product Suggestions