missing translation for 'onlineSavingsMsg'
Learn More

Brij(R) C20, Spectrum™ Chemical

Catalog No. 18601065
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601065 500 g Amber Glass Bottle
18601064 25 g Amber Glass Bottle
18601068 100 g Amber Glass Bottle
18601066 2.5 kg Amber Glass Bottle
18601067 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601065 Supplier Spectrum Chemical Mfg Cor Supplier No. BR106500GM
Only null left

BR106, 9004-95-9, C16H33(OCH2CH2)20OH

Spectrum™ Chemical Brij C20 is a non-ionic detergent for protein extraction and permeabilization of cells. It may be used in the preparation of yeast spheroplasts. It appears as a white, waxy solid.

Chemical Identifiers

CAS 9004-95-9
Molecular Formula C56H114O21
Molecular Weight (g/mol) 1123.51
MDL Number MFCD00080892
InChI Key NLMKTBGFQGKQEV-UHFFFAOYSA-N
IUPAC Name 3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxahexaheptacontan-1-ol
SMILES CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

Specifications

CAS Max % 100%
Quantity 500 g
Packaging Amber Glass Bottle
Grade Ungraded
Chemical Name or Material Brij 58