Learn More
Brady™ BBP™ Tamper-Evident Silver Metallized Polyester Label
BBP™ Tamper-Evident Silver Metallized Polyester Labels B-438 have a permanent adhesive that leaves a checkerboard footprint if removed.
$237.96 - $385.33
Specifications
Certifications/Compliance RoHS 2002/95/EC, UL969
Description Tamper-Evident Metallised Labels for BBP3x Printers
Adhesive Type Acrylic, Tamper-evident
Coating None
Color Silver
View More Specs
Products 4
Catalog Number Mfr. No. Height (English) Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Height (English) Length (English) Price Quantity & Availability  
19211101
View Documents Promotion Details
Brady™
B30C3000438SL
3 in. 100 ft.
Each for $304.03
Only null left
19211114
View Documents Promotion Details
Brady™
B3017438
2 in. 1 in.
Each for $237.96
Only null left
19211115
View Documents Promotion Details
Brady™
B3018438
3 in. 1 in.
Each for $362.35
Only null left
19211116
View Documents Promotion Details
Brady™
B30222438
3 in. 2 in.
Each for $385.33
Only null left
Description
Description
  • Designed for rating and serial plates requiring both high-performance and protection against removal
  • Ideal for product labeling, asset tracking and general industrial ID
  • Withstands solvents and variable temperatures
  • Requires minimal handling in order to prevent prematurely exposed checkerboard pattern
Specifications
Specifications
RoHS 2002/95/EC, UL969
Acrylic, Tamper-evident
Silver
Label
Thermal Transfer
For asset tracking, general ID
1
Brady Workstation, LabelMark, MarkWare
-40°C to +37°C
0.0033 in.
Tamper-Evident Metallised Labels for BBP3x Printers
None
Matte
Polyester
Non-sterile
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
1 Roll
-40°F to +100°F
60 lb./in.
0.084 mm