Learn More
Brady™ BBP™ Tamper-Evident Silver Metallized Polyester Label
BBP™ Tamper-Evident Silver Metallized Polyester Labels B-438 have a permanent adhesive that leaves a checkerboard footprint if removed.
Supplier:  Brady™ B3017438
 View more versions of this product
Catalog No. 19211114
Only null left
Description
Description
  • Designed for rating and serial plates requiring both high-performance and protection against removal
  • Ideal for product labeling, asset tracking and general industrial ID
  • Withstands solvents and variable temperatures
  • Requires minimal handling in order to prevent prematurely exposed checkerboard pattern
Specifications
Chemical Identifiers
RoHS 2002/95/EC, UL969
Acrylic, Tamper-evident
Silver
Label
Thermal Transfer
For asset tracking, general ID
1
Brady Workstation, LabelMark, MarkWare
-40°C to +37°C
0.0033 in.
Tamper-Evident Metallised Labels for BBP3x Printers
None
Matte
Polyester
Non-sterile
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
1 Roll
-40°F to +100°F
60 lb./in.
0.084 mm
Specifications
RoHS 2002/95/EC, UL969
Acrylic, Tamper-evident
Silver
Label
Single-side Printable
Label
For asset tracking, general ID
1
1225 Labels/Box
Rectangle
-40°F to +100°F
60 lb./in.
0.084 mm
2 x 1 in.
2.54 cm
25.4 mm
5.08 cm
Tamper-Evident Metallised Labels for BBP3x Printers
None
Matte
Polyester
Thermal Transfer
Non-sterile
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
-40°C
1 Roll
Brady Workstation, LabelMark, MarkWare
-40°C to +37°C
0.0033 in.
28.60 mm
1 in.
1 in.
2 in.
B30-17-438
Product Suggestions
Product Suggestions