Learn More
Brady™ BBP™ Hi-Performance Polyester Tape
BBP™ Hi-Performance Polyester Tape (B-569) has low halide content and is designed for marking stainless steel pipes particularly in harsh environments.
Supplier:  Brady™ B30C4000569YL
 Afficher plus de versions de ce produit
Numéro de catalogue. 19211239
Il en reste null
Description
Description
  • Durable, ultra-smooth polyester labels with permanent pressure sensitive acrylic adhesive
  • Outdoor grade polyester tape can be thermal transfer printed
  • Withstands mild chemicals, oil and water
  • Enables printers with XY cutters to cut custom shapes or multiple labels out of a single continuous roll
Spécifications
Identifiants chimiques
Acrylic, Permanent, Pressure-sensitive
Tape
Non-sterile
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
3 to 5 Years
-40 to 212°F
45 lb./in.
100 ft.
Gloss
Polyester
Designed for use on Stainless Steel pipes in harsh environments such as the pulp and Paper industry
80 Gardner Units
1 Roll
-40°C to +100°C
0.003 in.
30.48 m
Spécifications
High Performance Polyester Tape for BBP3x Printers
Yellow
Tape
Non-sterile
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
1
1 Roll
-40 to 212°F
45 lb./in.
0.076 mm
30.48 m
101 mm
Acrylic, Permanent, Pressure-sensitive
Gloss
Polyester
Designed for use on Stainless Steel pipes in harsh environments such as the pulp and Paper industry
80 Gardner Units
3 to 5 Years
Brady Workstation, LabelMark, MarkWare
-40°C to +100°C
0.003 in.
100 ft.
4 in.
B30C-4000-569-YL
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS