missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron nitride, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Melting Point | 2700.0°C to 3000.0°C |
| Quantity | 50 g |
| Assay Percent Range | 97.5% min |
| Linear Formula | BN |
| Merck Index | 15,1347 |
| Solubility Information | Solubility in water: slightly soluble (20°C). Other solubilities: soluble in water (95°) |
| IUPAC Name | boranylidyneamine |
| Formula Weight | 24.82 |
| Percent Purity | 98% |
| Chemical Name or Material | Boron nitride |
Sécurité et manipulation
| EINECSNumber | 233-136-6 |
RUO – Research Use Only