missing translation for 'onlineSavingsMsg'
Learn More

Bismuth AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Bi), LabChem™

Catalog No. lc116701
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC116701 500 mL Poly Bottle
LC116707 125 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC116701 Supplier LabChem Supplier No. LC116701
Only null left

Chemical Identifiers

CAS 10035-06-0 , 7697-37-2 , 7732-18-5
Molecular Formula BiH10N3O14
Molecular Weight (g/mol) 485.07
MDL Number MFCD00149157
InChI Key FBXVOTBTGXARNA-UHFFFAOYSA-N
PubChem CID 160911
IUPAC Name bismuth(3+) pentahydrate trinitrate
SMILES O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Poly Bottle
CAS Max % 95.92, 3.85, 0.23
Identification Passes Test
Viscosity 0.99cSt
Density 1.02g/mL
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg Bi)
Linear Formula Bi(NO2)3·5H2O
UN Number UN2031
Color Colorless
Solubility Soluble in water
Quantity 500 mL
Formula Weight 485.07
Grade Certified
Chemical Name or Material Bismuth AA Standard, 1000ppm (1mL = 1mg Bi)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature