missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Biotin-XX, SE (6-((6-((Biotinoyl)Amino)Hexanoyl)Amino)Hexanoic Acid, Succinimidyl Ester)

Catalog No. B1606
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1606 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1606 Supplier Invitrogen™ Supplier No. B1606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biotin-XX, SE (6-((6-((Biotinoyl)Amino)Hexanoyl)Amino)Hexanoic Acid, Succinimidyl Ester)

The amine-reactive biotin-XX, SE can be used to attach this important hapten to biomolecules of interest for subsequent detection with streptavidin, avidin or NeutrAvidin™ biotin-binding protein. This molecule has a 14 atom spacer designated by 'XX' to facilitate reactivity with the molecule and eventual accessibility of the biotin to avidin or streptavidin.

Specifications

Quantity 100 mg
Product Type Biotin-XX SE
Content And Storage Store in freezer (-5 to -30°C).
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Biotin-XX
Label Type Biotin and Other Haptens

For Research Use Only. Not for use in diagnostic procedures.