missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Biotin Alkyne (PEG4 carboxamide-Propargyl Biotin)

Catalog No. B10185
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10185 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B10185 Supplier Molecular Probes™ Supplier No. B10185

biotin alkyne (PEG4 carboxamide-propargyl biotin)

The hapten, biotin azide is reactive with terminal via a copper-catalyzed click reaction. Biotin can be subsequently detected with streptavidin, avidin or NeutrAvidin™ biotin-binding protein.

Specifications

Format Solid
Quantity 1 mg
Product Type Biotin Alkyne
Molecular Weight (g/mol) 528.66 Da
Solubility DMSO (Dimethylsulfoxide)
Content And Storage Store at ≤-20°C and keep desiccated.
Chemical Reactivity Azide
Reactive Group Alkyne
Shipping Condition Room Temperature
Reactive Moiety Alkyne
Label or Dye Biotin
Label Type Biotin and Other Haptens
Color None
Product Line Molecular Probes
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.