Learn More
Thermo Scientific™ BioLite™ Microwell Plates
Perform general cell culture procedures more economically with Thermo Scientific™ BioLite™ Cell Culture Treated Plates that are guaranteed to be sterile, non-pyrogenic, and non-cytotoxic.
$553.35 - $1,335.43
Specifications
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Color Clear
Sterility Sterile
Surface Treatment Cell Culture
View More Specs
Products 5
Catalog Number Mfr. No. No. of Wells Culture Area Volume (Metric) Well Price Quantity & Availability  
Catalog Number Mfr. No. No. of Wells Culture Area Volume (Metric) Well Price Quantity & Availability  
12556004
View Documents Promotion Details
Thermo Scientific™
130184
6 9.6 cm2 3 mL
Case of 50 Each for $795.57
Only null left
12556005
View Documents Promotion Details
Thermo Scientific™
130185
12 3.8 cm2 2 mL
Case of 50 Each for $888.93
Only null left
01549765
View Documents Promotion Details
Thermo Scientific™
130186
24 1.9 cm2 1 mL
Case of 50 Each for $553.35
Only null left
12556007
View Documents Promotion Details
Thermo Scientific™
130187
48 0.75 cm2 0.5 mL
Case of 50 Each for $1,221.79
Only null left
12556008
View Documents Promotion Details
Thermo Scientific™
130188
96 0.32 cm2 0.2 mL
Case of 50 Each for $1,335.43
Only null left
Description
Description
Experience optimal performance with wide selections in size, Thermo Scientific™ BioLite™ cell culture treated plates with lids suit a multitude of experimental needs while providing cost efficiency for a wide selection of applications in your cell culture routine.
• Sterile per ISO 11137, SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• BSE/TSE Free
• RNase/DNase Free
• USP Class VI raw material
Specifications
Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Clear
Sterile
Cell Culture
Videos
Safety and Handling
Safety and Handling
ShelfLife : 2 years