missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioLite™ Microwell Plates

Catalog No. 12556005
Change view
Click to view available options
No. of Wells:
12
24
48
6
96
Culture Area:
0.32 cm2
0.75 cm2
1.9 cm2
3.8 cm2
9.6 cm2
Volume (Metric) Well:
0.2 mL
0.5 mL
1 mL
2 mL
3 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells Culture Area Volume (Metric) Well
12556005 12 3.8 cm2 2 mL
12556004 6 9.6 cm2 3 mL
01549765 24 1.9 cm2 1 mL
12556007 48 0.75 cm2 0.5 mL
12556008 96 0.32 cm2 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 12556005 Supplier Thermo Scientific™ Supplier No. 130185
Season of Savings
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform general cell culture procedures more economically with Thermo Scientific™ BioLite™ Cell Culture Treated Plates that are guaranteed to be sterile, non-pyrogenic, and non-cytotoxic.

Experience optimal performance with wide selections in size, Thermo Scientific™ BioLite™ cell culture treated plates with lids suit a multitude of experimental needs while providing cost efficiency for a wide selection of applications in your cell culture routine.

• Sterile per ISO 11137, SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• BSE/TSE Free
• RNase/DNase Free
• USP Class VI raw material

Specifications

Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Lid Clear Closure Included
Culture Area 3.8 cm2
Array 3 x 4
Color Clear
Product Type Multi Dish
Volume (Metric) Well 2 mL
No. of Wells 12
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
No. per Case 50/Cs.
No. per Pack 1 (Individually Wrapped)
Show More Show Less
What is the recommended working volume of Nunc adherent cell culture products?

This depends largely on the cell line and working conditions (considerations include headspace for gas exchange, supplements used, and optimal feeding schedule, among others) but we do offer guidelines for development purposes (0.3 to 0.5 mL per cm2) . Note that these volumes are approximate and may have to be modified for best performance. T25 Flask = 7 mL, T75 Flask = 25 mL, T175 Flask = 55 mL, T225 Flask = 70 mL, 300cm2 Flask = 100 mL, 500 cm2 TripleFlask = 200 mL, 35 mm round culture dish = 3 mL, 60 mm round culture dish = 5 mL, 100 mm round culture dish = 12 mL, 150 mm round culture dish = 35 mL, 4 (rectangular) well Multidish = 5 mL, 8 (rectangular) well Multidish = 3 mL, 4 (round) well Multidish = 1 mL, 6 well Multidish = 3 mL, 12 well Multidish = 2 mL, 24 well Multidish = 1 mL, 48 well Multidish = 0.5 mL.