missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Biocytin (ε-Biotinoyl-L-Lysine)

Catalog No. B1592
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1592 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1592 Supplier Invitrogen™ Supplier No. B1592

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biocytin (ε-Biotinoyl-L-Lysine)

Biocytin can be used as microinjectable anterograde and transneuronal tracers that is fixable with aldehyde-based fixatives. Additionally the primary amine on the lysine can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a generate stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3). Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247).

Specifications

Product Type Biocytin
Quantity 100 mg
Content And Storage Store at room temperature.
Reagent Type Biocytins
Shipping Condition Room Temperature
Label Type Biotin & Other Haptens

For Research Use Only. Not for use in diagnostic procedures.