Learn More
Current Technologies Bio-Screen™ Biohazard Wipes, Liners, and Rolls
Low-linting, white, absorbent wipes bonded to an orange plastic backing
Supplier:  Current Technologies BH44985
 Afficher plus de versions de ce produit
Numéro de catalogue. 1441252E
Il en reste null
Description
Description
  • White absorbent side of this wipe / liner is low-linting and more absorbent than gauze or paper wipes
  • Orange plastic backing of wipe / liner blocks soak through of blood, body fluids, specimens and chemicals (xylene, ketones, formalin) to gloves and surfaces
  • Used for cleaning up small / large spills, to wipe pipets, probes, slide prep devices, and to pop caps from blood and specimen tubes
  • Limit cross-contamination from gloves as employees touch phones, keyboards, doors, etc.
  • Larger sizes can be used as mats and liners. Rolls of material can be cut to desired sizes
  • Three thicknesses available: Standard (absorbs 9 times its weight), Heavy (absorbs 12 times its weight), Super (absorbs 25 times its weight)
Spécifications
Identifiants chimiques
White and Orange
Used for cleaning up small or large spills, to wipe pipets, probes, slide prep devices, and to pop caps from blood and specimen tubes. Wipes and Liners are non-sterile and not intended for use on patients.
Each
Spécifications
Biohazard Wipe
4 in.
4 in.
4 x 4 in. (101.6 x 101.6mm)
Used for cleaning up small or large spills, to wipe pipets, probes, slide prep devices, and to pop caps from blood and specimen tubes. Wipes and Liners are non-sterile and not intended for use on patients.
Standard
101.6 mm
101.6 mm
White and Orange
Each
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
Wipes and Liners are non-sterile and not intended for use on patients.