missing translation for 'onlineSavingsMsg'
Learn More

Current Technologies Bio-Screen™ Biohazard Wipes, Liners, and Rolls

Catalog No. p-210864
Change view
Click to view available options
Dimensions (L x W):
100 ft. x 16 in.
16 x 11 in. (406.4 x 279.4mm)
16 x 16 in. (406.4 x 406.4mm)
3 x 3 in. (76.2 x 76.2mm)
4 x 4 in. (101.6 x 101.6mm)
9 x 8 in. (228.6 x 203.2mm)
Absorbancy:
High
Standard
Super
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Absorbancy
1451252H 3 x 3 in. (76.2 x 76.2mm) Super
1441252D 4 x 4 in. (101.6 x 101.6mm) High
1441257 16 x 11 in. (406.4 x 279.4mm) High
1441252C 3 x 3 in. (76.2 x 76.2mm) Standard
1441252E 4 x 4 in. (101.6 x 101.6mm) Standard
144124 100 ft. x 16 in. Super
144123 9 x 8 in. (228.6 x 203.2mm) Super
1441252B 100 ft. x 16 in. Standard
1441258 16 x 16 in. (406.4 x 406.4mm) High
1441252G 4 x 4 in. (101.6 x 101.6mm) Super
1441252A 100 ft. x 16 in. High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 1451252H Supplier Current Technologies Supplier No. BH91033
Only null left

Low-linting, white, absorbent wipes bonded to an orange plastic backing

  • White absorbent side of this wipe / liner is low-linting and more absorbent than gauze or paper wipes
  • Orange plastic backing of wipe / liner blocks soak through of blood, body fluids, specimens and chemicals (xylene, ketones, formalin) to gloves and surfaces
  • Used for cleaning up small / large spills, to wipe pipets, probes, slide prep devices, and to pop caps from blood and specimen tubes
  • Limit cross-contamination from gloves as employees touch phones, keyboards, doors, etc.
  • Larger sizes can be used as mats and liners. Rolls of material can be cut to desired sizes
  • Three thicknesses available: Standard (absorbs 9 times its weight), Heavy (absorbs 12 times its weight), Super (absorbs 25 times its weight)

Specifications

Product Type Biohazard Wipe
Absorbancy Super
Length (English) 3 in.
Length (Metric) 76.2 mm
Width (English) 3 in.
Width (Metric) 76.2 mm
Dimensions (L x W) 3 x 3 in. (76.2 x 76.2mm)
Color White and Orange
For Use With (Application) Used for cleaning up small or large spills, to wipe pipets, probes, slide prep devices, and to pop caps from blood and specimen tubes. Wipes and Liners are non-sterile and not intended for use on patients.
Quantity Each

Wipes and Liners are non-sterile and not intended for use on patients.