missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ BigDye XTerminator™ Purification Kit

The BigDye XTerminator™ Purification Kit is a fast, simple purification method for DNA sequencing reactions that removes unincorporated BigDye™ terminators and salts.

$538.00 - $43,840.00

Specifications

Format Kit
Sample Type Sequencing Products
Final Product Type DNA
For Use With (Application) Sequencing
Includes 2 Bottles
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
4376486
Encompass
View Documents Promotion Details
Applied Biosystems™
4376486
100 Preps
Each for $538.00
Only null left
 
4376487
Encompass
View Documents Promotion Details
Applied Biosystems™
4376487
1000 Preps
Each for $4,435.00
Only null left
 
4376484
Encompass
View Documents Promotion Details
Applied Biosystems™
4376484
2500 Preps
Each for $7,240.00
Only null left
 
4376485
Encompass
View Documents Promotion Details
Applied Biosystems™
4376485
40,000 Preps N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Eliminate liquid transfer - Use a single plate
  • Obtain complete dye blob removal
  • Stabilize the samples before analysis
  • Improve your sequencing workflow
  • Perform manual or automated protocols
  • Get rapid, reliable, and reproducible results

Order Info

Each

Specifications

Specifications

Kit
DNA
2 Bottles
Sequencing Reactions
Sequencing Products
Sequencing
BigDye XTerminator, BigDye
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.