missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ BigClean™-Lysis Matrix Tube with 50 Stainless Steel Balls
BigClean™-Lysis Matrix Tube with 50 Stainless Steel Balls use to break hard/brittle samples
$504.63 - $1,520.21
Spécifications
| Content And Storage | Room temperature |
|---|---|
| Sample Type | Animal Tissue, Bacteria (Gram +/-), Bone, Feces, Fixed Samples, Food, Fungi, Heart, Lung, Plants, Roots, Seeds, Skeletal Muscle, Sludge, Soil, Spores, Wastewater, Yeast |
| Matrix | Stainless-steel |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|
116941010
|
MP Biomedicals™
116941010 |
10 x 50 mL |
paquet for $504.63
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
MP116941050
|
MP Biomedicals™
116941050 |
50 x 50 mL |
chaque for $1,520.21
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
- Specialty applications include RNA extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing
- Each impact-resistant 50 mL tube contains fifty (50) 1/4 in. (6.3 mm, 0.25 cal.) stainless-steel grinding balls
Certifications
DNase and RNase free
Spécifications
| Room temperature | |
| Animal Tissue, Bacteria (Gram +/-), Bone, Feces, Fixed Samples, Food, Fungi, Heart, Lung, Plants, Roots, Seeds, Skeletal Muscle, Sludge, Soil, Spores, Wastewater, Yeast | |
| Stainless-steel |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.