missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BetaBasic™ Phenyl Columns, 5μm Particle Size

Catalog No. 71805254630
Change view
Click to view available options
Length (Metric):
150 mm
250 mm
Diameter (Metric):
4.6 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
71805254630 250 mm 4.6 mm
71805154630 150 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71805254630 Supplier Thermo Scientific™ Supplier No. 71805254630

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BetaBasic columns are highly efficient and reproducible for small molecules, peptides and protein digests

  • 150Å pore size for small molecules, peptides and protein digests
  • Superb pH stability

Order Info

Other column dimensions are also available. Please call Customer Service for more information.

Specifications

Column Format Analytical Column
Diameter (Metric) 4.6 mm
Film Thickness 5 μm
For Use With LC/MS Separation
Functionality/Interaction Type Stable bonding
Length (Metric) 250 mm
Particle Shape Spherical
Particle Size 5 μm
Pore Size 150 Å
Surface Area 200 m²/g
USP Type L11
Stationary Phase Phenyl (Ph)
Product Line Beta
Packing Material Silica
Carbon Load 7%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Product Type Column
Show More Show Less