missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Hydroxyisovaleric Acid 96.0+%, TCI America™
Supplier: TCI America H070125G
| Quantity | 25 g |
|---|
Identifiants chimiques
| C5H10O3 | |
| MFCD00059081 | |
| beta-hydroxyisovaleric acid, 3-hydroxyisovaleric acid, 3-hydroxy-3-methylbutyric acid, hmb-d6, 3-oh-isovaleric acid, 3-hydroxy-isovaleric acid, 3-hydroxy-3-methyl-butanoic acid, butanoic acid, 3-hydroxy-3-methyl, b-hydroxyisovaleric acid, beta-hydroxy-beta-methylbutyrate | |
| CHEBI:37084 | |
| CC(C)(O)CC(O)=O |
Spécifications
| beta-Hydroxyisovaleric Acid | |
| Yellow | |
| 25 g | |
| beta-hydroxyisovaleric acid, 3-hydroxyisovaleric acid, 3-hydroxy-3-methylbutyric acid, hmb-d6, 3-oh-isovaleric acid, 3-hydroxy-isovaleric acid, 3-hydroxy-3-methyl-butanoic acid, butanoic acid, 3-hydroxy-3-methyl, b-hydroxyisovaleric acid, beta-hydroxy-beta-methylbutyrate | |
| CC(C)(O)CC(O)=O | |
| 118.13 | |
| CHEBI:37084 | |
| ≥96.0% (GC,T) |
| 625-08-1 | |
| C5H10O3 | |
| MFCD00059081 | |
| AXFYFNCPONWUHW-UHFFFAOYSA-N | |
| 3-hydroxy-3-methylbutanoic acid | |
| 69362 | |
| 118.13 | |
| Liquid |
Sécurité et manipulation
EINECSNumber : (2)-1429
TSCA : No