missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Hydroxyisovaleric Acid 96.0+%, TCI America™
Chemical Identifiers
| CAS | 625-08-1 |
|---|---|
| Molecular Formula | C5H10O3 |
| Molecular Weight (g/mol) | 118.13 |
| MDL Number | MFCD00059081 |
| InChI Key | AXFYFNCPONWUHW-UHFFFAOYSA-N |
| Synonym | beta-hydroxyisovaleric acid, 3-hydroxyisovaleric acid, 3-hydroxy-3-methylbutyric acid, hmb-d6, 3-oh-isovaleric acid, 3-hydroxy-isovaleric acid, 3-hydroxy-3-methyl-butanoic acid, butanoic acid, 3-hydroxy-3-methyl, b-hydroxyisovaleric acid, beta-hydroxy-beta-methylbutyrate |
| PubChem CID | 69362 |
| ChEBI | CHEBI:37084 |
| IUPAC Name | 3-hydroxy-3-methylbutanoic acid |
| SMILES | CC(C)(O)CC(O)=O |
Chemical Identifiers
| C5H10O3 | |
| MFCD00059081 | |
| beta-hydroxyisovaleric acid, 3-hydroxyisovaleric acid, 3-hydroxy-3-methylbutyric acid, hmb-d6, 3-oh-isovaleric acid, 3-hydroxy-isovaleric acid, 3-hydroxy-3-methyl-butanoic acid, butanoic acid, 3-hydroxy-3-methyl, b-hydroxyisovaleric acid, beta-hydroxy-beta-methylbutyrate | |
| CHEBI:37084 | |
| CC(C)(O)CC(O)=O |
Specifications
| 625-08-1 | |
| C5H10O3 | |
| beta-hydroxyisovaleric acid, 3-hydroxyisovaleric acid, 3-hydroxy-3-methylbutyric acid, hmb-d6, 3-oh-isovaleric acid, 3-hydroxy-isovaleric acid, 3-hydroxy-3-methyl-butanoic acid, butanoic acid, 3-hydroxy-3-methyl, b-hydroxyisovaleric acid, beta-hydroxy-beta-methylbutyrate | |
| CC(C)(O)CC(O)=O | |
| 118.13 | |
| CHEBI:37084 | |
| ≥96.0% (GC,T) | |
| beta-Hydroxyisovaleric Acid |
| Yellow | |
| MFCD00059081 | |
| AXFYFNCPONWUHW-UHFFFAOYSA-N | |
| 3-hydroxy-3-methylbutanoic acid | |
| 69362 | |
| 118.13 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1429
TSCA : No