missing translation for 'onlineSavingsMsg'
Learn More

Beryllium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLBE22X

 View more versions of this product

Catalog No. PLBE2-2X

Only null left

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Be1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Beryllium Standard
7440-41-7 , 7697-37-2 , 7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,2.0,97.9
MFCD00134032
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
ATBAMAFKBVZNFJ-UHFFFAOYSA-N
2% HNO3
9.01
5460467
Assurance
Triple-leached LDPE bottle
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
0.1,2.0,97.9
Be
AA, ICP
Certified Reference Material
[Be]
beryllium
500 mL
CHEBI:33783
Matrix    Concentration    Cat. No.
2-5% HNO31000μg/mL    PLBE2-2Y, -2X    10,000    PLBE2-3Y, -3X

Chemical Identifiers

7440-41-7
9.01
ATBAMAFKBVZNFJ-UHFFFAOYSA-N
CHEBI:33783
[Be]
Be
MFCD00134032
5460467
beryllium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

May cause cancer. Causes severe skin burns and eye damage.

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only