missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzalkonium Chloride Solution, NF, 47.5-52.5%, Spectrum™ Chemical
B1068, 8001-54-5
$791.46 - $23810.04
Chemical Identifiers
| CAS | 8001-54-5 |
|---|---|
| Molecular Formula | C9H13ClN |
| Molecular Weight (g/mol) | 170.66 |
| SMILES | [Cl-].C[N+](C)(*)CC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18600875
|
Spectrum Chemical Mfg Cor
B1068500MLGL |
500 mL | Amber Glass Bottle |
Each for $791.46
|
|
||||
|
18600876
|
Spectrum Chemical Mfg Cor
B10684LTGL |
4 L | Amber Glass Bottle |
Each for $2,695.75
|
|
||||
|
18600877
|
Spectrum Chemical Mfg Cor
B106820LTBL |
20 L | Amber Glass Bottle |
Each for $5,379.14
|
|
||||
|
18600878
|
Spectrum Chemical Mfg Cor
B1068200LTBL |
200 L | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Benzalkonium Chloride Solution, 50 Percent, NF is used in the pharmaceutical industry as an antimicrobial preservative, antiseptic, disinfectant, solubilizing agent, and wetting agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.