missing translation for 'onlineSavingsMsg'
Learn More

Benzalkonium Chloride Solution, NF, 47.5-52.5%, Spectrum™ Chemical

Catalog No. 18600876
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600876 4 L Amber Glass Bottle
18600875 500 mL Amber Glass Bottle
18600877 20 L Amber Glass Bottle
18600878 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600876 Supplier Spectrum Chemical Mfg Cor Supplier No. B10684LTGL
Only null left

B1068, 8001-54-5

Spectrum™ Chemical Benzalkonium Chloride Solution, 50 Percent, NF is used in the pharmaceutical industry as an antimicrobial preservative, antiseptic, disinfectant, solubilizing agent, and wetting agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8001-54-5 , 7732-18-5 , 64-17-5 , 100-44-7
Molecular Formula C9H13ClN
Molecular Weight (g/mol) 170.66
SMILES [Cl-].C[N+](C)(*)CC1=CC=CC=C1

Specifications

CAS Max % 50%, 46%, 4%, ≤0.05%
Concentration 50%
Quantity 4 L
Packaging Amber Glass Bottle
UN Number UN2920
Percent Purity 47.5 to 52.5%
Grade NF
Chemical Name or Material Benzalkonium Chloride Solution