accessibility menu, dialog, popup

UserName

NH3CCH3RgpCl

CAS RN 8001-54-5

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 170.66
Molecular Formula: C9H13ClN
InChi Key: —
SMILES: [Cl-].C[N+](C)(*)CC1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (3)
  • (3)
Grade 
  • (3)
  • (1)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (14)
  • (9)
  • (1)
  • (9)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)