missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Polypropylene Sterilizing Trays

Corrosion-proof polypropylene plastic trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.

$95.75 - $199.07

Specifications

Color White
Height (Metric) 12.7 cm
Material Polypropylene
Type Sterilizing Tray
Height (English) 5 in.
View More Specs

Products 3
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
22087114
Accessories View Documents Promotion Details
Bel Art™
H16260 0000
5 L
Each for $95.75
Case of 6 Each for $574.50
Only null left
 
22087189
Accessories View Documents Promotion Details
Bel Art™
H16264 0000
18 L
Each for $199.07
Case of 6 Each for $1,194.42
Only null left
 
1525216
Accessories View Documents Promotion Details
Bel Art™
H162620000
13 L
Each for $166.04
Case of 6 Each for $996.24
Only null left
 
Description

Description

Corrosion-proof polypropylene plastic trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.

  • Ideal for sterilizing instruments, bottles containing agar, deionized water, saline and other reagents, and for safely cleaning glassware in sinks
  • Trays are steam autoclavable at 121°C (250°F)
  • Covers are sold separately

Order Info

Covers are sold separately.

Specifications

Specifications

White
Polypropylene
5 in.
Autoclavable
12.7 cm
Sterilizing Tray
Rectangular
Videos
SDS
Documents

Documents