missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ Polypropylene Sterilizing Trays
Description
Corrosion-proof polypropylene plastic trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.
- Ideal for sterilizing instruments, bottles containing agar, deionized water, saline and other reagents, and for safely cleaning glassware in sinks
- Trays are steam autoclavable at 121°C (250°F)
- Covers are sold separately
Order Info
Covers are sold separately.
Specifications
Specifications
| Capacity (Metric) | 13 L |
| Dimensions (L x W x H) | 19 x 10.5 x 5 in. (48.3 x 26.7 x 12.7 cm) |
| Color | White |
| Height (Metric) | 12.7 cm |
| Length (Metric) | 48.3 cm |
| Material | Polypropylene |
| Type | Sterilizing Tray |
| Width (Metric) | 26.7 cm |
| For Use With | Fits 13L Tray Cover, Mfr. No. H162610000 |
| Length (English) | 19 in. |
| Show More |