missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ Polypropylene Sterilizing Trays
Description
Corrosion-proof polypropylene plastic trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.
- Ideal for sterilizing instruments, bottles containing agar, deionized water, saline and other reagents, and for safely cleaning glassware in sinks
- Trays are steam autoclavable at 121°C (250°F)
- Covers are sold separately
Order Info
Covers are sold separately.
Specifications
Specifications
| Capacity (Metric) | 5 L |
| Dimensions (L x W x H) | 12 x 8 x 5 in. (30.5 x 20.3 x 12.7 cm) |
| Color | White |
| Height (Metric) | 12.7 cm |
| Length (Metric) | 30.5 cm |
| Material | Polypropylene |
| Type | Sterilizing Tray |
| Width (Metric) | 20.3 cm |
| For Use With | Fits 5L Tray Cover, Mfr. No. H162590000 |
| Length (English) | 12 in. |
| Show More |