missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Resurge™ CD Pak
GREENER_CHOICE

Catalog No. 01801000
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01801000 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01801000 Supplier Gibco™ Supplier No. 670030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Resurge CD Pak are a diverse set of chemically defined supplement formulations to complement a variety of base media. The CD PAK contains one of each Resurge supplement: CD1, CD2, CD3, CD4, and CD5 at 100 grams each.

Resurge CD Pak are a diverse set of chemically defined supplement formulations to complement a variety of base media. The CD PAK contains one of each Resurge supplement: CD1, CD2, CD3, CD4, and CD5 at 100 grams each. The supplements have been developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. In order to increase versatility of the formulation, glucose and L-glutamine are not included in the final formulations. Therefore, these two components should be added by users and monitored as appropriate.

Specifications

Content And Storage •One of each Resurge supplement: CD1, CD2, CD3, CD4, and CD5 at 100 grams each.
•Storage conditions: 2° to 8°C.
•Store in cool, dry conditions in well-sealed containers.
Cell Line CHO
Classification Animal Origin-free, Chemically-defined, Protein-free
Form Powder
Product Type Media Supplement
Format Bottles
Product Line Resurge
Quantity 1 Ea.
Source Animal origin-free