missing translation for 'onlineSavingsMsg'
Learn More

Dunlop™ Bata Viking Galoshes and Overboots

Numéro de catalogue. p-37985
Change view
Click to view available options
Size:
Small
Medium
Large
2X-Large
For Use With (Equipment):
Size 10 to 11 shoes
Size 14 to 15 shoes
Size 6 to 7 shoes
Size 8 to 9 shoes
Height (English):
4 in.
17 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Size For Use With (Equipment) Height (English)
17986360 Medium Size 8 to 9 shoes 4 in.
189991319A Medium Size 8 to 9 shoes 17 in.
189991319B Large Size 10 to 11 shoes 17 in.
19051925 Small Size 6 to 7 shoes 17 in.
19051931 2X-Large Size 14 to 15 shoes 17 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Numéro de catalogue. 17986360 Fournisseur Dunlop™ Code fournisseur 86010MD00
Il en reste null

Spécifications

Color Black
For Use With (Application) Used in construction, farming, utilities and food processing
For Use With (Equipment) Size 8 to 9 shoes
Height (English) 4 in.
Lug Type Kick-off
Material Flexothane, PVC
Size Medium
Sole Features 4-way cleated, slip-resistant outsole
Toe Style Plain
Waterproof Yes