missing translation for 'onlineSavingsMsg'
Learn More

Barium ICP Standard, 10,000 ppm Ba in 5% HNO3, Ricca Chemical

$1097.70 - $1097.70

Chemical Identifiers

CAS 10022-31-8
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
PubChem CID 24798
IUPAC Name barium(2+) dinitrate
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PBA10KN-500
View Documents
Ricca Chemical Company
PBA10KN500
500 mL Natural Poly Bottle
Each for $1,097.70
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10022-31-8
261.34
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
barium(2+) dinitrate
BaN2O6
MFCD00003442
24798
[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Natural Poly Bottle
10,000ppm Ba
93.0,7.0,1.78
Liquid
MFCD00003442
< 2
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
barium(2+) dinitrate
500 mL
Barium ICP Standard
10022-31-8 , 7697-37-2 , 7732-18-5
90.0,6.73,1.71
1 mL = 10 mg Ba
BaN2O6
Colorless
Standard
[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
261.34
24798
Videos
SDS
Documents

Documents