missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium ICP Standard, 1000 ppm Ba in 3% HNO3, Ricca Chemical
$317.30 - $317.30
Chemical Identifiers
| CAS | 10022-31-8 |
|---|---|
| Molecular Formula | BaN2O6 |
| Molecular Weight (g/mol) | 261.34 |
| MDL Number | MFCD00003442 |
| InChI Key | IWOUKMZUPDVPGQ-UHFFFAOYSA-N |
| PubChem CID | 24798 |
| IUPAC Name | barium(2+) dinitrate |
| SMILES | [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PBA1KN-100
|
Ricca Chemical Company
PBA1KN100 |
100 mL | Natural LDPE Bottle |
Each for $317.30
|
|
||||
Chemical Identifiers
| 10022-31-8 | |
| 261.34 | |
| IWOUKMZUPDVPGQ-UHFFFAOYSA-N | |
| barium(2+) dinitrate |
| BaN2O6 | |
| MFCD00003442 | |
| 24798 | |
| [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
| Natural LDPE Bottle | |
| 1,000ppm Ba | |
| 97.0,4.39,0.19 | |
| Liquid | |
| MFCD00003442 | |
| < 2 | |
| Standard | |
| [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 261.34 | |
| 24798 |
| 10022-31-8 , 7697-37-2 , 7732-18-5 | |
| 94.0,4.22,0.18 | |
| 1 mL = 1 mg Ba | |
| BaN2O6 | |
| Colorless | |
| Approximately 0°C | |
| IWOUKMZUPDVPGQ-UHFFFAOYSA-N | |
| barium(2+) dinitrate | |
| 100 mL | |
| Barium ICP Standard |