missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, 0.100 N (N/10), 0.0500 M (M/20), Ricca Chemical

$118.07 - $377.77

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
88016
View Documents
Ricca Chemical Company
88016
500 mL Natural Poly Bottle
Each for $118.07
Only null left
 
880-32
View Documents
Ricca Chemical Company
88032
1 L Natural Poly Bottle
Each for $201.50
Only null left
 
8801
View Documents
Ricca Chemical Company
RI8801
4 L Natural Poly Bottle
Each for $377.77
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10326-27-9
244.26
PWHCIQQGOQTFAE-UHFFFAOYSA-L
5284346
barium(2+) dihydrate dichloride
BaCl2H4O2
MFCD00149154
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
CHEBI:86153
O.O.[Cl-].[Cl-].[Ba++]

Specifications

Colorless
97.0,1.18
BaCl2H4O2
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
O.O.[Cl-].[Cl-].[Ba++]
244.26
CHEBI:86153
Natural Poly Bottle
500 mL
0.100Normal,0.0500Molar
10326-27-9 , 7732-18-5
100.0,1.23
MFCD00149154
PWHCIQQGOQTFAE-UHFFFAOYSA-L
barium(2+) dihydrate dichloride
5284346
Laboratory
∼5 to 7
Liquid
Barium Chloride
Videos
SDS
Documents

Documents