missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride, 0.100 N (N/10), 0.0500 M (M/20), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 97.0, 1.18 |
| CAS Max % | 100.0, 1.23 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| pH | ∼5 to 7 |
| Quantity | 1 L |
| Physical Form | Liquid |
| Concentration or Composition Notes | 0.100Normal,0.0500Molar |
| Chemical Name or Material | Barium Chloride |