missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, 0.100 N (N/10), 0.0500 M (M/20), Ricca Chemical

Supplier:  Ricca Chemical Company RI8801

 View more versions of this product

Catalog No. 8801


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Barium Chloride
10326-27-9 , 7732-18-5
100.0,1.23
MFCD00149154
PWHCIQQGOQTFAE-UHFFFAOYSA-L
barium(2+) dihydrate dichloride
5284346
Laboratory
∼5 to 7
Liquid
Colorless
97.0,1.18
BaCl2H4O2
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
O.O.[Cl-].[Cl-].[Ba++]
244.26
CHEBI:86153
Natural Poly Bottle
4 L
0.100Normal,0.0500Molar

Chemical Identifiers

10326-27-9
244.26
PWHCIQQGOQTFAE-UHFFFAOYSA-L
5284346
barium(2+) dihydrate dichloride
BaCl2H4O2
MFCD00149154
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
CHEBI:86153
O.O.[Cl-].[Cl-].[Ba++]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions