Learn More
Motic BA310E Microscope and Camera Bundle
The BA310 is designed for the rigors of daily routine work in the demanding applications of universities, clinics, laboratories, and many other life sciences or medical applications requiring quality optical performance.
Supplier:  Motic 1100402700082
 Afficher plus de versions de ce produit
Numéro de catalogue. 01257482
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Description
Description
  • These models introduce a new rackless stage concept without a prominent gear rack to allow for a more convenient use of the X/Y movement.
  • Allow easy interchangeability of its halogen bulb with LED modules
  • Swiveling, 30° inclined Siedentopf type
  • Reversed quintuple revolving nosepiece
  • Coaxial coarse and fine focusing adjustment with tension adjustment
  • Built in low position coaxial rackless stage (Right-hand control)
  • Diopter Adjustment: ±5
Spécifications
Identifiants chimiques
BA310E LED Trinocular
Spécifications
BTI10 Camera Bundle
CCIS™ EC Plan Achromat Objectives EC PL (4X, 10X, 40X(S), 100X(S-oil))
Trinocular
LED
10X
BA310E LED Trinocular
Moticam BTI10
WF 10X/20 mm
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS