Learn More
Motic BA210S LED Compound Microscope
Numéro de catalogue. 01257472
Change view
Click to view available options
Head Style:
Binocular
Trinocular
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Head Style
01257472 Binocular
01257473 Trinocular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 01257472 Fournisseur Motic Code fournisseur 1100100403831
Il en reste null
The BA210S biological compound microscope is designed for advanced biology students, teachers, doctors, and lab technicians.
  • Every aspect of these microscopes contributes to making your daily work easier.
  • The ergonomic shape of the stand and the comfortability of the different heads enables fatigue-free viewing for many hours.
Spécifications
Magnification Power 10X
Objective Types 4X/0.10 (DT 17mm), 10X/0.25 (DT 6.4mm), 40X/0.65/S (DT 0.45mm), 100X/1.25/S (DT 0.14mm)
Head Style Binocular
Lighting Type LED
Eyepiece WF 10X/20 mm