Learn More
United Scientific™ B.O.D. Bottles, Numbered
Efficiently conduct the 5-day biochemical oxygen demand (BOD) test with United Scientific™ BOD Bottles, designed for incubating diluted samples of clean, waste, effluent, and polluted water. Feature a barcode for streamlined tracking and glass stoppers for secure sealing.
Supplier:  United Scientific™ BOD300N97
 Afficher plus de versions de ce produit
Numéro de catalogue. S012433
Il en reste null
Description
Description
  • Bottles are numbered to allow for convenient organization of samples
  • 300 mL bottles also include a barcode
  • Glass robotic stoppers are included
  • 300 mL line is available in a series of numbers from 1 through 120 in increments of 24
  • Made of borosilicate glass
  • Easy-to-read numbers
  • Barcode for easy recording
  • Write on marking spaces
Spécifications
Identifiants chimiques
Glass Robotic
Spécifications
300 mL
97 to 120
Glass Robotic
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS