missing translation for 'onlineSavingsMsg'
Learn More

Aztreonam

Catalog No. AAJ6288706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6288706 5 g
AAJ6288703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6288706 Supplier Thermo Scientific Chemicals Supplier No. J6288706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 78110-38-0 | C13H17N5O8S2 | 435.426 g/mol

Aztreonam is used to treat severe infections of the urinary tract, lower respiratory tract, skin, stomach, female reproductive organs, and other body systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aztreonam is used to treat severe infections of the urinary tract, lower respiratory tract, skin, stomach, female reproductive organs, and other body systems.

Solubility
Soluble in DMF/water (1:1) at 50 mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 78110-38-0
Molecular Formula C13H17N5O8S2
Molecular Weight (g/mol) 435.426
MDL Number MFCD00072145
InChI Key WZPBZJONDBGPKJ-PSGLRMFWSA-N
PubChem CID 56842327
IUPAC Name 2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid
SMILES CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N

Specifications

Type Aztreonam
Physical Form Powder
Melting Point 227°C (decomposition)
Color White
Quantity 5 g
Merck Index 14,925
Solubility Information Soluble in DMF/water (1:1) at 50 mg/ml
Formula Weight 435.43
Chemical Name or Material Aztreonam
EINECSNumber 278-839-9
RTECSNumber UA2451400
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only