Learn More
Axygen™ Tip Refill System
Numéro de catalogue. 14222436
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 200 μL
15 to 1250 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Volume (Metric)
14222436 15 to 1250 μL
14222437 1 to 200 μL
14222438 0.1 to 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 14222436 Fournisseur Axygen™ Code fournisseur RFL1200C
Il en reste null
Axygen™ style and universal fit pipet tips in easy-to-load refill racks
Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.
  • Certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Translucent rack allows tips to be easily seen
  • Color-coded insert allows for easy identification of tip
  • Spécifications
    Volume (Metric) 15 to 1250 μL
    Sterility Non-sterile
    Color Clear
    Autoclavable Autoclavable
    Format Rack
    Tip Style Universal
    Packaging 96 Tips/Rack, 8 Racks/Pk, 6 Packs/Cs
    Graduated No