missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human AURKC (Aurora C), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
AURKC (Aurora C) is a chromosomal passenger protein that forms complexes with AURKB (Aurora B) and inner centromere proteins, playing a role in organizing microtubules in relation to centrosome/spindle function during mitosis. It is overexpressed in several cancer cell lines, suggesting an involvement in oncogenic signal transduction. Studies on the temporal expression pattern and subcellular localization of Aurora kinases in mitotic cells suggest an association with mitotic structure. Aurora kinase functional influences span from G2 phase to cytokinesis and may be involved in key cell cycle events such as centrosome duplication, chromosome bi-orientation and segregation, cleavage furrow positioning, and ingression.
Spécifications
Spécifications
| Accession Number | Q9UQB9 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 6795 |
| Molecular Weight (g/mol) | 61.7 kDa |
| Name | Human AURKC (Aurora C), GST Tag |
| Quantity | 10 μg |
| Regulatory Status | For research use only. Not for use in diagnostic procedures |
| Gene Alias | AIE1; AIE2; AIK3; AIRK3; ARK3; ARK-3; AurC; AURKC; Aurora 3; aurora B; aurora kinase C; aurora/Ipl1/Eg2 protein 1; aurora/IPL1/EG2 protein 2; aurora/IPL1-related kinase 3; aurora-C; aurora-related kinase 3; epididymis secretory protein Li 90; HEL-S-90; IAK3; serine/threonine kinase 13 (aurora/IPL1-like); serine/threonine kinase 13 (aurora/IPL-like); serine/threonine-protein kinase 13; Serine/threonine-protein kinase aurora-C; SPGF5; STK13 |
| Afficher plus de résultats |