Learn More
3M™ N95 Aura™ Particulate Respirators, 9205+
Catalog No. 19072522
Change view
Click to view available options
Quantity:
240/Cs.
440/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19072522 240/Cs.
296368418 440/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19072522 Supplier 3M™ Supplier No. 7100232219
Three-panel, flat-fold disposable respirators provide comfortable, reliable worker protection against non-oil based particles.
  • Provides reliable protection of at least 95 percent filtration efficiency against certain non-oil based particles
  • Lightweight, three-panel designed respirator provides quality, reliable, and convenient worker protection
  • Suggested applications: Grinding, sanding, sweeping, bagging and other dusty or arid operations
  • Used to help reduce inhalation of certain airborne biological particles like mold, Bacillus anthracis and Mycobacterium tuberculosis
  • Example applications include emergency or pandemic preparedness planning, stockpiling, etc.
Specifications
Aerosol Type N - Not Resistant to Oil
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Design Type Flat-fold
Disposable Single-use
Filter Type N95
Product Type General Purpose Particulate Respirator
Size Standard
With Exhalation Valve No
Filter Efficiency 95%
Model 9205+
Nosepiece Type Adjustable Nose Clip
Quantity 240/Cs.
Sterility Non-sterile
Strap Attachment Type Stapled Headbands
FDA Cleared Surgical Mask No
Show More Show Less