missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Astec™ CHIROBIOTIC™ T Chiral HPLC Column, 5 μm Particle Size
CHIROBIOTIC™ T columns utilize teicoplanin as a chiral selector.
$2308.53 - $13124.76
Specifications
| Particle Size | 5 μm |
|---|---|
| pH | 3.8 to 6.8 |
| Pore Size | 100 Å |
| Stationary Phase | Teicoplanin Glycopeptide |
| Product Line | Astec CHIROBIOTIC |
| Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity | ||||
|
111006050
|
MilliporeSigma™ Supelco™
12018AST |
10 cm | 2.1 mm |
Each for $3,720.60
|
|
||||
|
111005913
|
MilliporeSigma™ Supelco™
12019AST |
15 cm | 2.1 mm |
Each for $4,308.32
|
|
||||
|
111005863
|
MilliporeSigma™ Supelco™
12020AST |
25 cm | 2.1 mm |
Each for $5,224.49
|
|
||||
|
111006182
|
MilliporeSigma™ Supelco™
12010AST |
10 cm | 3 mm |
Each for $3,342.90
|
|
||||
|
111006529
|
MilliporeSigma™ Supelco™
12021AST |
5 cm | 4.6 mm |
Each for $2,308.53
|
|
||||
|
111006200
|
MilliporeSigma™ Supelco™
12022AST |
10 cm | 4.6 mm |
Each for $3,538.25
|
|
||||
|
111005984
|
MilliporeSigma™ Supelco™
12023AST |
15 cm | 4.6 mm |
Each for $4,241.05
|
|
||||
|
111005874
|
MilliporeSigma™ Supelco™
12024AST |
25 cm | 4.6 mm |
Each for $5,205.40
|
|
||||
|
111005634
|
MilliporeSigma™ Supelco™
12034AST |
25 cm | 10 mm |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
CHIROBIOTIC™ T columns offer unique selectivity for a number of classes of molecules, specifically underivatized alpha, beta, gamma and cyclic amino acids, N-derivatized amino acids, hydroxy-carboxylic acids, acidic compounds including carboxylic acids and phenols, small peptides, neutral aromatic analytes and cyclic aromatic and aliphatic amines. Separations normally obtained on a chiral crown ether or ligand exchange-type CSPs are also possible on the CHIROBIOTIC™ T and T2, but in much simpler mobile phases, like water-alcohol. In addition, all of the known beta-blockers (amino alcohols), and dihydrocoumarins have been resolved. CHIROBIOTIC™ T and T2 differ in their bonding chemistry and the pore size of the support particle, giving them different selectivity and preparative capacity.Specifications
| 5 μm | |
| 100 Å | |
| Astec CHIROBIOTIC | |
| Chiral |
| 3.8 to 6.8 | |
| Teicoplanin Glycopeptide | |
| Teicoplanin Glycopeptide Phase/High-Purity Silica Gel Particle Platform | |
| Chiral HPLC |
Astec and CHIROBIOTIC are registered trademarks of Sigma-Aldrich Co. LLC.