missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Astec™ CHIROBIOTIC™ T Chiral HPLC Column, 5 μm Particle Size
Description
CHIROBIOTIC™ T columns offer unique selectivity for a number of classes of molecules, specifically underivatized alpha, beta, gamma and cyclic amino acids, N-derivatized amino acids, hydroxy-carboxylic acids, acidic compounds including carboxylic acids and phenols, small peptides, neutral aromatic analytes and cyclic aromatic and aliphatic amines. Separations normally obtained on a chiral crown ether or ligand exchange-type CSPs are also possible on the CHIROBIOTIC™ T and T2, but in much simpler mobile phases, like water-alcohol. In addition, all of the known beta-blockers (amino alcohols), and dihydrocoumarins have been resolved. CHIROBIOTIC™ T and T2 differ in their bonding chemistry and the pore size of the support particle, giving them different selectivity and preparative capacity.
Specifications
Specifications
| Diameter (Metric) | 2.1 mm |
| Length (Metric) | 25 cm |
| Particle Size | 5 μm |
| pH | 3.8 to 6.8 |
| Pore Size | 100 Å |
| Stationary Phase | Teicoplanin Glycopeptide |
| Product Line | Astec CHIROBIOTIC |
| Matrix | Teicoplanin Glycopeptide Phase/High-Purity Silica Gel Particle Platform |
| Phase | Chiral |
| Technique | Chiral HPLC |
Astec and CHIROBIOTIC are registered trademarks of Sigma-Aldrich Co. LLC.