missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASPM Monoclonal Antibody (K52002_11G4), Invitrogen™
Mouse Monoclonal Antibody
$1,601.06 - $2,821.92
Spécifications
| Antigen | ASPM |
|---|---|
| Clone | K52002_11G4 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|
MA556234
|
Invitrogen™
MA556234 |
200 μL |
chaque for $2,821.92
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
MA555854
|
Invitrogen™
MA555854 |
50 μL |
chaque for $1,601.06
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Sequence of this protein is as follows: CKVPLSNENS QVPQSPEDWR KSEVSPRIPE CQGSKSPKAI FEELVEMKSN YYSFIKQNNP KFSAVQDISS HSHNKQPKRR PILSATVTKR KATCTRENQT EINKPKAKRC LNSAVGEHEK VINNQKEKED FHSYLPIIDP ILSKSKSYKN EVTPSSTTAS VARKRKSDGS MEDANVRVAI TEHTEVREIK RIHFSPSEPK TSAVKKTKNV TTPISKRISN REKLNLKKKT DLSIFRTPIS KTNKRTKPII AVAQSSLTFI KPLKTDIPRH PMPFAAKNMF YDERWKEKQE QGFTWWLNFI LTPDDFTVKT
ASPM Monoclonal antibody specifically detects ASPM in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpécifications
| ASPM | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| abnormal spindle microtubule assembly; Abnormal spindle protein homolog; Abnormal spindle-like microcephaly-associated protein; ASP; asp (abnormal spindle) homolog, microcephaly associated; Asp homolog; ASPM; Calmbp1; MCPH5 | |
| Recombinant human ASPM (immunogen range is 411-720). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| ASPM |
| K52002_11G4 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8IZT6 | |
| 259266 | |
| IgG2b | |
| Protein A | |
| Antibody |