missing translation for 'onlineSavingsMsg'
Learn More

Arsenite Standard, 0.100 N (N/10), Ricca Chemical

Catalog No. 82016
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
82016 500 mL Natural Poly Bottle
82032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 82016 Supplier Ricca Chemical Company Supplier No. RI82016

Chemical Identifiers

CAS 1327-53-3 , 144-55-8 , 1310-73-2 , 7732-18-5
Molecular Formula As2O3
Molecular Weight (g/mol) 197.84
MDL Number MFCD00003433
InChI Key IKWTVSLWAPBBKU-UHFFFAOYSA-N
IUPAC Name diarsorosooxidane
SMILES O=[As]O[As]=O

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 0.100Normal
CAS Min % 97.0, 0.48, 0.39, 0.21
CAS Max % 100.0, 0.5, 0.4, 0.22
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 500 mL
Chemical Name or Material Arsenite Standard