missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arsenic ICP Standard, 1000 ppm As in 3% HNO3, Ricca Chemical
$266.12 - $266.12
Chemical Identifiers
| CAS | 7440-38-2 |
|---|---|
| Molecular Formula | As |
| Molecular Weight (g/mol) | 74.92 |
| MDL Number | MFCD00085309 |
| InChI Key | RQNWIZPPADIBDY-UHFFFAOYSA-N |
| PubChem CID | 5359596 |
| ChEBI | CHEBI:27563 |
| IUPAC Name | arsenic |
| SMILES | [As] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PAS1KN-100
|
Ricca Chemical Company
PAS1KN100 |
100 mL | Natural LDPE Bottle |
Each for $266.12
|
|
||||
Chemical Identifiers
| 7440-38-2 | |
| 74.92 | |
| RQNWIZPPADIBDY-UHFFFAOYSA-N | |
| CHEBI:27563 | |
| [As] |
| As | |
| MFCD00085309 | |
| 5359596 | |
| arsenic |
Specifications
| Natural LDPE Bottle | |
| 1,000ppm As | |
| 94.0,7.78,0.09 | |
| Liquid | |
| MFCD00085309 | |
| < 2 | |
| Standard | |
| [As] | |
| 74.92 | |
| 5359596 | |
| Arsenic ICP Standard |