missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARP (N-(aminooxyacetyl)-N'-(D-Biotinoyl) hydrazine, Trifluoroacetic Acid Salt)

Catalog No. A10550
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10550 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10550 Supplier Invitrogen™ Supplier No. A10550

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ARP (N-(aminooxyacetyl)-N'-(D-Biotinoyl) hydrazine, Trifluoroacetic Acid Salt)

ARP (Aldehyde-Reactive Probe) is a biotinylated hydroxylamine that reacts with the exposed aldehyde group formed at abasic sites in damaged DNA. The biotins bound to the abasic sites can then be quantitated with fluorescent or enzyme-conjugated streptavidin complexes. ARP is suitable for use in microplate assays.

Specifications

Quantity 10 mg
Product Type ARP
Content And Storage Store in freezer (-5 to -30°C).
Chemical Reactivity Carboxylic Acid, Ketone, Aldehyde
Shipping Condition Room Temperature
Reactive Moiety Amine, Hydroxylamine
Label or Dye Biotin
Label Type Biotin and Other Haptens

For Research Use Only. Not for use in diagnostic procedures.