missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 20 cal Arc Flash Kit with Voltage Gloves (10)

Numéro de catalogue. p-7220167
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Size
170011296 Small
170011301 Medium
170011306 Large
170011311 X-Large
170011316 2X-Large
170011321 3X-Large
170011326 4X-Large
170011331 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Numéro de catalogue. 170011296 Fournisseur National Safety Apparel™ Code fournisseur KIT2SC20SM10
Il en reste null

Arc flash kit with PureView™ FR jacket and bib overall

  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • FR hook and loop front closure on short coat
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Hook and loop at side of bib for easy access
  • Expandable leg opening to fit over boots
  • Made in the USA

Spécifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Product Type Arc Flash Kit
Certifications/Compliance CAT 2, NFPA 70E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Gender Unisex
Arc Flash Rating 20 cal/cm²
Ankle Style Open
Seam Type Stitched
Size Small
Closure Type FR Hook and Loop
Color Navy
Suspender Adjustable
Material Westex™ UltraSoft™
Afficher plus de résultats Afficher moins de résultats