Learn More
Fisherbrand™ Drosophila Vials
Straight wall design minimizes the dislodging of food; provides more efficient racking, filling and plugging.
 Afficher plus de versions de ce produit
Numéro de catalogue. AS510
Il en reste null
Description
Description
Straight wall design minimizes the dislodging of food; provides more efficient racking, filling and plugging
Fisherbrand Drosophila vials are available in clear polystyrene, translucent and flexible polypropylene, and virtually unbreakable K-resin. These vials feature a flat bottom, straight wall design for convenient racking, filling and stacking. Available bulk packaged or in a convenient, 10 × 10 tray pack configuration.
Polypropylene Vials
  • Translucent and flexible
  • Autoclavable
Polystyrene Vials
  • Excellent clarity for optimal visibility
  • Nonautoclavable
K-Resin Vials
  • Highly flexible, virtually unbreakable
  • Nonautoclavable
Standard-Packed Glass Shell Vials
  • Type 1, N-51A borosilicate glass
  • Securely packed in cardboard trays with individual vial dividers
Bulk- and Tray-Packed Shell Vials
  • Available in narrow- or wide-mouth styles
  • Constructed from polystyrene, K-Resin™, or polypropylene
  • K-Resin and polypropylene vials are virtually unbreakable, scratch and mar resistant; polystyrene and K-Resin vials have glass-like clarity
  • Polypropylene vials are autoclavable; polystyrene and K-Resin are nonautoclavable
Order Info
Bulk-packed vials are layered 500 per case and oriented with all open ends facing one way. Tray-packed vials are shrink-wrapped in a 10 x 10 in. (25 x 25cm) row configuration with dividers between vials.
Spécifications
Identifiants chimiques
Clear
Straight wall design minimizes the dislodging of food, provides more efficient racking, filling and plugging
3.74 in.
95 mm
Spécifications
3.74 in.
29.21 mm
Straight wall design minimizes the dislodging of food, provides more efficient racking, filling and plugging
Not Autoclavable
Clear
1.15 in.
95 mm
Wide
K-Resin
Drosophila Vial, Tray Packed
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Accessoires