missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Universal Master Mix II, with UNG

TaqMan Universal Master Mix II is a convenient mix of components (except primers, probes, template, and water) necessary to perform a real-time polymerase chain reaction (PCR).

$203.00 - $8,800.00

Specifications

Concentration 2X
Content And Storage Store at 2°C to 8°C in the dark.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
Detection Method Primer-probe
GC-Rich PCR Performance High
PCR Method qPCR
View More Specs

Requires: Primers, probes, template, water


Products 6
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
4440042
Encompass
View Documents Promotion Details
Applied Biosystems™
4440042
1 x 1 mL
Each for $203.00
Only null left
 
4440038
Encompass
View Documents Promotion Details
Applied Biosystems™
4440038
1 x 5 mL
Each for $938.00
Only null left
 
4440044
Encompass
View Documents Promotion Details
Applied Biosystems™
4440044
2 x 5 mL
Each for $1,882.00
Only null left
 
4440045
Encompass
View Documents Promotion Details
Applied Biosystems™
4440045
5 x 5 mL
Each for $4,505.00
Only null left
 
4440046
Encompass
View Documents Promotion Details
Applied Biosystems™
4440046
10 x 5 mL
Each for $8,800.00
Only null left
 
4440039
Encompass
View Documents Promotion Details
Applied Biosystems™
4440039
1 x 50 mL
Each for $8,430.00
Only null left
 
Description

Description

TaqMan Universal Master Mix II is a convenient mix of components (except primers, probes, template, and water) necessary to perform a real-time polymerase chain reaction (PCR). Use TaqMan Universal Master Mix II to amplify complementary DNA (cDNA) and DNA targets for a variety of applications, including quantitation and genotyping.

This TaqMan Universal Master Mix II is supplied as a 2X concentrate and contains:
• AmpliTaq Gold DNA Polymerase, UP (Ultra Pure)
• dNTPs (with dUTP)
• ROX Passive Reference
• Uracil-N-glycosylase (UNG)
• Optimized buffer components

Note: This TaqMan Universal Master Mix II contains uracil-N-glycoslyase (UNG).

TaqMan Universal Master Mix II has been optimized for use with primers and TaqMan probes that have been designed according to Thermo Fisher Scientific guidelines. The master mix can be used with custom TaqMan assays available from our custom assay service, or with pre-optimized assays such as:
• TaqMan gene expression assays
• TaqMan microRNA assays
• TaqMan drug metabolism genotyping assays
• TaqMan SNP genotyping assays

For RNA quantitation experiments, TaqMan Universal Master Mix II is used in the second step of a two-step reverse transcription/polymerase chain reaction (RT-PCR) protocol. The cDNA template used with the master mix can be generated in a reverse transcription reaction using cDNA synthesis kits available from Thermo Fisher Scientific.

Alternative product: Try TaqMan Fast Advanced Master Mix (Cat. No. 4444963), our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of TaqMan Universal Master Mix II and added additional capabilities for your gene expression analysis.

Order Info

Guaranteed minimum shelf life is 60 days. Shipping Conditions: Wet Ice.

Specifications

Specifications

2X
Primer-probe
qPCR
Standard
ROX (Pre-mixed)
Universal Master Mix II
Gene Expression, Genotyping, miRNA analysis
DNA (Genomic), cDNA
Store at 2°C to 8°C in the dark.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
High
AmpliTaq Gold DNA Polymerase
7300 System, 7500 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System
TaqMan
UP (Ultra Pure)
2 X
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.