missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Universal Master Mix II, with UNG

TaqMan Universal Master Mix II is a convenient mix of components (except primers, probes, template, and water) necessary to perform a real-time polymerase chain reaction (PCR).

Supplier:  Applied Biosystems™ 4440045

Encompass

 View more versions of this product

Catalog No. 4440045


Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Only null left

Requires : Primers, probes, template, water

Description

Description

TaqMan Universal Master Mix II is a convenient mix of components (except primers, probes, template, and water) necessary to perform a real-time polymerase chain reaction (PCR). Use TaqMan Universal Master Mix II to amplify complementary DNA (cDNA) and DNA targets for a variety of applications, including quantitation and genotyping.

This TaqMan Universal Master Mix II is supplied as a 2X concentrate and contains:
• AmpliTaq Gold DNA Polymerase, UP (Ultra Pure)
• dNTPs (with dUTP)
• ROX Passive Reference
• Uracil-N-glycosylase (UNG)
• Optimized buffer components

Note: This TaqMan Universal Master Mix II contains uracil-N-glycoslyase (UNG).

TaqMan Universal Master Mix II has been optimized for use with primers and TaqMan probes that have been designed according to Thermo Fisher Scientific guidelines. The master mix can be used with custom TaqMan assays available from our custom assay service, or with pre-optimized assays such as:
• TaqMan gene expression assays
• TaqMan microRNA assays
• TaqMan drug metabolism genotyping assays
• TaqMan SNP genotyping assays

For RNA quantitation experiments, TaqMan Universal Master Mix II is used in the second step of a two-step reverse transcription/polymerase chain reaction (RT-PCR) protocol. The cDNA template used with the master mix can be generated in a reverse transcription reaction using cDNA synthesis kits available from Thermo Fisher Scientific.

Alternative product: Try TaqMan Fast Advanced Master Mix (Cat. No. 4444963), our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of TaqMan Universal Master Mix II and added additional capabilities for your gene expression analysis.

Order Info

Guaranteed minimum shelf life is 60 days. Shipping Conditions: Wet Ice.

Specifications

Chemical Identifiers

2X
Primer-probe
qPCR
Standard
ROX (Pre-mixed)
Universal Master Mix II
Gene Expression, Genotyping, miRNA analysis
DNA (Genomic), cDNA
Store at 2°C to 8°C in the dark.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
High
AmpliTaq Gold DNA Polymerase
7300 System, 7500 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System
TaqMan
UP (Ultra Pure)
2 X

Specifications

2X
Primer-probe
qPCR
Standard
ROX (Pre-mixed)
Universal Master Mix II
5 x 5 mL
2 X
55 mL
Store at 2°C to 8°C in the dark.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
High
AmpliTaq Gold DNA Polymerase
7300 System, 7500 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System
TaqMan
UP (Ultra Pure)
Gene Expression, Genotyping, miRNA analysis
DNA (Genomic), cDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.